Assay Details
| Gene Symbol: Aga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503041.2 | XP_003503089.1 | 3 - 4 | 498 | 67 | |
| GenBank mRNA | JI879076.1 | - | 3 - 4 | 469 | 67 | |
| JP040099.1 | - | 3 - 4 | 469 | 67 | ||
More Information
Set Membership: |
|

Aga
Genomic Map