Assay Details
| Gene Symbol: Asb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506650.1 | XP_003506698.1 | 2 - 3 | 482 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asb4| Gene Symbol: Asb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506650.1 | XP_003506698.1 | 2 - 3 | 482 | 64 | |
Set Membership: |
|