Assay Details
| Gene Symbol: Dscr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507265.2 | XP_003507313.2 | 2 - 3 | 399 | 58 | |
| GenBank mRNA | JI869112.1 | - | 2 - 3 | 399 | 58 | |
More Information
Set Membership: |
|

Dscr3
Genomic Map