Assay Details
| Gene Symbol: Gpr97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509802.1 | XP_003509850.1 | 5 - 6 | 610 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr97| Gene Symbol: Gpr97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509802.1 | XP_003509850.1 | 5 - 6 | 610 | 75 | |
Set Membership: |
|