Assay Details
| Gene Symbol: Ndufs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509919.2 | XP_003509967.1 | 5 - 6 | 604 | 58 | |
| XM_007650564.1 | XP_007648754.1 | 4 - 5 | 421 | 58 | ||
| XM_007650565.1 | XP_007648755.1 | 6 - 7 | 769 | 58 | ||
| XM_007650566.1 | XP_007648756.1 | 6 - 7 | 811 | 58 | ||
| XM_007650567.1 | XP_007648757.1 | 6 - 7 | 596 | 58 | ||
| XM_007650568.1 | XP_007648758.1 | 5 - 6 | 420 | 58 | ||
| GenBank mRNA | JI873291.1 | - | 4 - 5 | 370 | 58 | |
More Information
Set Membership: |
|

Ndufs8
Genomic Map