Assay Details
| Gene Symbol: Ca5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511521.2 | XP_003511569.2 | 3 - 4 | 335 | 67 | |
| XM_007652076.1 | XP_007650266.1 | 3 - 4 | 297 | 67 | ||
| XM_007652077.1 | XP_007650267.1 | 2 - 3 | 298 | 67 | ||
| XM_007652078.1 | XP_007650268.1 | 2 - 3 | 213 | 67 | ||
| XM_007652079.1 | XP_007650269.1 | 2 - 3 | 211 | 67 | ||
| GenBank mRNA | JP048144.1 | - | 2 - 3 | 248 | 67 | |
More Information
Set Membership: |
|

Ca5b
Genomic Map