Assay Details
| Gene Symbol: Sae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649605.1 | XP_007647795.1 | 4 - 5 | 545 | 68 | |
| GenBank mRNA | JP058916.1 | - | 4 - 5 | 477 | 68 | |
More Information
Set Membership: |
|

Sae1
Genomic Map