Assay Details
| Gene Symbol: Ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647729.1 | XP_007645919.1 | 4 - 5 | 465 | 69 | |
| XM_007647730.1 | XP_007645920.1 | 5 - 6 | 462 | 69 | ||
| XM_007647731.1 | XP_007645921.1 | 6 - 7 | 546 | 69 | ||
| XM_007647732.1 | XP_007645922.1 | 6 - 7 | 525 | 69 | ||
More Information
Set Membership: |
|

Ak1
Genomic Map