Assay Details
| Gene Symbol: Kbtbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497324.2 | XP_003497372.1 | 2 - 3 | 779 | 75 | |
| XM_003497325.2 | XP_003497373.1 | 2 - 3 | 679 | 75 | ||
More Information
Set Membership: |
|

Kbtbd4
Genomic Map