Assay Details
| Gene Symbol: Eef1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502109.2 | XP_003502157.1 | 1 - 2 | 414 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eef1b2| Gene Symbol: Eef1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502109.2 | XP_003502157.1 | 1 - 2 | 414 | 58 | |
Set Membership: |
|