Assay Details
| Gene Symbol: Rexo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502576.2 | XP_003502624.1 | 14 - 15 | 3670 | 64 | |
| XM_007643203.1 | XP_007641393.1 | 14 - 15 | 3796 | 64 | ||
| GenBank mRNA | JI874944.1 | - | 6 - 7 | 657 | 64 | |
| JP054698.1 | - | 6 - 7 | 657 | 64 | ||
More Information
Set Membership: |
|

Rexo1
Genomic Map