Assay Details
| Gene Symbol: Pcyt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505692.2 | XP_003505740.1 | 7 - 8 | 979 | 67 | |
| XM_007646162.1 | XP_007644352.1 | 7 - 8 | 1003 | 67 | ||
| XM_007646163.1 | XP_007644353.1 | 7 - 8 | 971 | 67 | ||
| GenBank mRNA | JP048718.1 | - | 6 - 7 | 715 | 67 | |
More Information
Set Membership: |
|

Pcyt1b
Genomic Map