Assay Details
| Gene Symbol: Ndufb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505967.2 | XP_003506015.1 | 3 - 4 | 461 | 77 | |
| XM_007646459.1 | XP_007644649.1 | 3 - 4 | 404 | 77 | ||
| XM_007646460.1 | XP_007644650.1 | 2 - 3 | 311 | 77 | ||
| GenBank mRNA | JI870935.1 | - | 3 - 4 | 443 | 77 | |
| JP051200.1 | - | 3 - 4 | 443 | 77 | ||
More Information
Set Membership: |
|

Ndufb6
Genomic Map