Assay Details
| Gene Symbol: Fgf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510065.1 | XP_003510113.1 | 2 - 3 | 143 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf4| Gene Symbol: Fgf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510065.1 | XP_003510113.1 | 2 - 3 | 143 | 103 | |
Set Membership: |
|