Assay Details
| Gene Symbol: Gabarapl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510161.2 | XP_003510209.1 | 3 - 4 | 322 | 75 | |
| GenBank mRNA | JI889706.1 | - | 2 - 3 | 207 | 75 | |
| JP046837.1 | - | 2 - 3 | 207 | 75 | ||
More Information
Set Membership: |
|

Gabarapl2
Genomic Map