Assay Details
| Gene Symbol: Lin7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510698.2 | XP_003510746.1 | 3 - 4 | 277 | 61 | |
| GenBank mRNA | JP042038.1 | - | 3 - 4 | 265 | 61 | |
More Information
Set Membership: |
|

Lin7c
Genomic Map