Assay Details
| Gene Symbol: Lrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513992.2 | XP_003514040.1 | 2 - 3 | 373 | 70 | |
| GenBank mRNA | JI867829.1 | - | 1 - 2 | 99 | 70 | |
More Information
Set Membership: |
|

Lrr1
Genomic Map