Assay Details
| Gene Symbol: Csdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514312.2 | XP_003514360.2 | 2 - 3 | 426 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csdc2| Gene Symbol: Csdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514312.2 | XP_003514360.2 | 2 - 3 | 426 | 59 | |
Set Membership: |
|