Assay Details
| Gene Symbol: Igsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653611.1 | XP_007651801.1 | 2 - 3 | 697 | 64 | |
| XM_007653612.1 | XP_007651802.1 | 2 - 3 | 697 | 64 | ||
More Information
Set Membership: |
|

Igsf6
Genomic Map