Assay Details
| Gene Symbol: Cd55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007616464.1 | XP_007614654.1 | 2 - 3 | 366 | 73 | |
| XM_007616471.1 | XP_007614661.1 | 2 - 3 | 366 | 73 | ||
| XM_007616474.1 | XP_007614664.1 | 2 - 3 | 366 | 73 | ||
| XM_007616483.1 | XP_007614673.1 | 2 - 3 | 366 | 73 | ||
| XM_007616494.1 | XP_007614684.1 | 2 - 3 | 366 | 73 | ||
| XM_007616503.1 | XP_007614693.1 | 2 - 3 | 366 | 73 | ||
| XM_007616509.1 | XP_007614699.1 | 2 - 3 | 366 | 73 | ||
More Information
Set Membership: |
|

Cd55
Genomic Map