Assay Details
| Gene Symbol: LOC100752965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652689.1 | XP_007650879.1 | 29 - 30 | 4822 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100752965| Gene Symbol: LOC100752965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652689.1 | XP_007650879.1 | 29 - 30 | 4822 | 103 | |
Set Membership: |
|