Assay Details
| Gene Symbol: LOC100752965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652689.1 | XP_007650879.1 | 55 - 56 | 9641 | 113 | |
| GenBank mRNA | JP061555.1 | - | 2 - 3 | 216 | 113 | |
More Information
Set Membership: |
|

LOC100752965
Genomic Map