Assay Details
| Gene Symbol: Shq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495203.2 | XP_003495251.1 | 10 - 11 | 1271 | 61 | |
| GenBank mRNA | JI873276.1 | - | 7 - 8 | 745 | 61 | |
| JP038174.1 | - | 7 - 8 | 745 | 61 | ||
More Information
Set Membership: |
|

Shq1
Genomic Map