Assay Details
| Gene Symbol: Rfesd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500004.2 | XP_003500052.1 | 2 - 3 | 872 | 113 | |
| XM_007640743.1 | XP_007638933.1 | 2 - 3 | 967 | 113 | ||
| XM_007640744.1 | XP_007638934.1 | 2 - 3 | 743 | 113 | ||
| GenBank mRNA | JI886946.1 | - | 1 - 2 | 176 | 113 | |
| JP064987.1 | - | 1 - 2 | 176 | 113 | ||
More Information
Set Membership: |
|

Rfesd
Genomic Map