Assay Details
| Gene Symbol: Rfesd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500004.2 | XP_003500052.1 | 3 - 4 | 910 | 103 | |
| XM_007640743.1 | XP_007638933.1 | 3 - 4 | 1005 | 103 | ||
| XM_007640744.1 | XP_007638934.1 | 3 - 4 | 781 | 103 | ||
| GenBank mRNA | JI886946.1 | - | 2 - 3 | 214 | 103 | |
| JP064987.1 | - | 2 - 3 | 214 | 103 | ||
More Information
Set Membership: |
|

Rfesd
Genomic Map