Assay Details
| Gene Symbol: Aldh5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502206.2 | XP_003502254.1 | 3 - 4 | 300 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh5a1| Gene Symbol: Aldh5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502206.2 | XP_003502254.1 | 3 - 4 | 300 | 54 | |
Set Membership: |
|