Assay Details
| Gene Symbol: Htatip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502264.2 | XP_003502312.1 | 3 - 4 | 462 | 64 | |
| GenBank mRNA | JI876555.1 | - | 2 - 3 | 342 | 64 | |
| JP053412.1 | - | 2 - 3 | 342 | 64 | ||
More Information
Set Membership: |
|

Htatip2
Genomic Map