Assay Details
| Gene Symbol: Hsp90b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505850.2 | XP_003505898.1 | 11 - 12 | 1525 | 64 | |
| GenBank mRNA | JI890704.1 | - | 11 - 12 | 1503 | 64 | |
| JP053748.1 | - | 11 - 12 | 1503 | 64 | ||
More Information
Set Membership: |
|

Hsp90b1
Genomic Map