Assay Details
| Gene Symbol: LOC100770272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506268.2 | XP_003506316.1 | 15 - 16 | 4129 | 79 | |
| XM_007646759.1 | XP_007644949.1 | 15 - 16 | 4091 | 79 | ||
More Information
Set Membership: |
|

LOC100770272
Genomic Map