Assay Details
| Gene Symbol: Unc93b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509920.2 | XP_003509968.2 | 7 - 8 | 959 | 62 | |
| GenBank mRNA | JI866431.1 | - | 7 - 8 | 921 | 62 | |
More Information
Set Membership: |
|

Unc93b1
Genomic Map