Assay Details
| Gene Symbol: LOC100763162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607746.1 | XP_007605936.1 | 1 - 2 | 194 | 95 | |
| GenBank mRNA | JP041678.1 | - | 1 - 2 | 194 | 95 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100763162| Gene Symbol: LOC100763162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607746.1 | XP_007605936.1 | 1 - 2 | 194 | 95 | |
| GenBank mRNA | JP041678.1 | - | 1 - 2 | 194 | 95 | |