Assay Details
| Gene Symbol: LOC100764372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646372.1 | XP_007644562.1 | 1 - 2 | 89 | 91 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100764372| Gene Symbol: LOC100764372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646372.1 | XP_007644562.1 | 1 - 2 | 89 | 91 | |