Assay Details
| Gene Symbol: Gbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654339.1 | XP_007652529.1 | 8 - 9 | 1571 | 63 | |
| XM_007654340.1 | XP_007652530.1 | 8 - 9 | 1571 | 63 | ||
More Information
Set Membership: |
|

Gbp5
Genomic Map