Assay Details
| Gene Symbol: Meaf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642863.1 | XP_007641053.1 | 1 - 2 | 227 | 64 | |
| GenBank mRNA | JP053918.1 | - | 1 - 2 | 227 | 64 | |
More Information
Set Membership: |
|

Meaf6
Genomic Map