Assay Details
| Gene Symbol: Ctla4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497416.1 | XP_003497464.1 | 1 - 2 | 109 | 77 | |
| XM_007611963.1 | XP_007610153.1 | 1 - 2 | 109 | 77 | ||
More Information
Set Membership: |
|

Ctla4
Genomic Map