Assay Details
| Gene Symbol: Scnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498667.2 | XP_003498715.1 | 6 - 7 | 703 | 62 | |
| GenBank mRNA | JI863698.1 | - | 6 - 7 | 703 | 62 | |
| JP057693.1 | - | 6 - 7 | 703 | 62 | ||
More Information
Set Membership: |
|

Scnm1
Genomic Map