Assay Details
| Gene Symbol: Gpr56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509803.2 | XP_003509851.1 | 13 - 14 | 1981 | 74 | |
| XM_007650448.1 | XP_007648638.1 | 13 - 14 | 2209 | 74 | ||
| XM_007650449.1 | XP_007648639.1 | 13 - 14 | 2264 | 74 | ||
| XM_007650450.1 | XP_007648640.1 | 13 - 14 | 2053 | 74 | ||
| XM_007650451.1 | XP_007648641.1 | 13 - 14 | 2012 | 74 | ||
| XM_007650452.1 | XP_007648642.1 | 13 - 14 | 1992 | 74 | ||
| XM_007650453.1 | XP_007648643.1 | 13 - 14 | 2194 | 74 | ||
| XM_007650454.1 | XP_007648644.1 | 13 - 14 | 2248 | 74 | ||
| GenBank mRNA | JI870114.1 | - | 11 - 12 | 1870 | 74 | |
| JP062986.1 | - | 12 - 13 | 1957 | 74 | ||
More Information
Set Membership: |
|

Gpr56
Genomic Map