Assay Details
| Gene Symbol: Gpr56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509803.2 | XP_003509851.1 | 2 - 3 | 111 | 100 | |
| XM_007650453.1 | XP_007648643.1 | 2 - 3 | 324 | 100 | ||
| XM_007650454.1 | XP_007648644.1 | 2 - 3 | 378 | 100 | ||
| GenBank mRNA | JP062986.1 | - | 1 - 2 | 87 | 100 | |
More Information
Set Membership: |
|

Gpr56
Genomic Map