Assay Details
| Gene Symbol: Gpr56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509803.2 | XP_003509851.1 | 4 - 5 | 670 | 75 | |
| XM_007650448.1 | XP_007648638.1 | 4 - 5 | 898 | 75 | ||
| XM_007650449.1 | XP_007648639.1 | 4 - 5 | 953 | 75 | ||
| XM_007650450.1 | XP_007648640.1 | 4 - 5 | 742 | 75 | ||
| XM_007650451.1 | XP_007648641.1 | 4 - 5 | 701 | 75 | ||
| XM_007650452.1 | XP_007648642.1 | 4 - 5 | 681 | 75 | ||
| XM_007650453.1 | XP_007648643.1 | 4 - 5 | 883 | 75 | ||
| XM_007650454.1 | XP_007648644.1 | 4 - 5 | 937 | 75 | ||
| GenBank mRNA | JI870114.1 | - | 2 - 3 | 559 | 75 | |
| JP062986.1 | - | 3 - 4 | 646 | 75 | ||
More Information
Set Membership: |
|

Gpr56
Genomic Map