Assay Details
| Gene Symbol: LOC100758683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510927.1 | XP_003510975.1 | 1 - 2 | 299 | 97 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100758683| Gene Symbol: LOC100758683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510927.1 | XP_003510975.1 | 1 - 2 | 299 | 97 | |