Assay Details
| Gene Symbol: LOC100758683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510927.1 | XP_003510975.1 | 7 - 8 | 1243 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100758683| Gene Symbol: LOC100758683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510927.1 | XP_003510975.1 | 7 - 8 | 1243 | 59 | |
Set Membership: |
|