Assay Details
| Gene Symbol: Cyr61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512942.2 | XP_003512990.2 | 4 - 5 | 1183 | 77 | |
| GenBank mRNA | JI883912.1 | - | 1 - 2 | 202 | 77 | |
| JP060801.1 | - | 3 - 4 | 757 | 77 | ||
More Information
Set Membership: |
|

Cyr61
Genomic Map