Assay Details
| Gene Symbol: Lcmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514320.2 | XP_003514368.1 | 8 - 9 | 899 | 65 | |
| GenBank mRNA | JI889181.1 | - | 8 - 9 | 912 | 65 | |
| JP042268.1 | - | 8 - 9 | 912 | 65 | ||
More Information
Set Membership: |
|

Lcmt1
Genomic Map