Assay Details
| Gene Symbol: Ahnak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007633200.1 | XP_007631390.1 | 6 | 2385 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ahnak2| Gene Symbol: Ahnak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007633200.1 | XP_007631390.1 | 6 | 2385 | 55 | |
Set Membership: |
|