Assay Details
| Gene Symbol: Tmem79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502913.2 | XP_003502961.1 | 2 - 3 | 1080 | 60 | |
| XM_007643530.1 | XP_007641720.1 | 3 - 4 | 1109 | 60 | ||
More Information
Set Membership: |
|

Tmem79
Genomic Map