Assay Details
| Gene Symbol: LOC100758957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506639.2 | XP_003506687.1 | 54 - 55 | 8702 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100758957| Gene Symbol: LOC100758957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506639.2 | XP_003506687.1 | 54 - 55 | 8702 | 97 | |
Set Membership: |
|