Assay Details
| Gene Symbol: Ankrd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509237.2 | XP_003509285.1 | 5 - 6 | 950 | 75 | |
| XM_007649825.1 | XP_007648015.1 | 3 - 4 | 4919 | 75 | ||
| XM_007649826.1 | XP_007648016.1 | 3 - 4 | 1350 | 75 | ||
| GenBank mRNA | JP055033.1 | - | 2 - 3 | 329 | 75 | |
More Information
Set Membership: |
|

Ankrd10
Genomic Map