Assay Details
| Gene Symbol: Ovol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653323.1 | XP_007651513.1 | 8 - 9 | 1533 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ovol1| Gene Symbol: Ovol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653323.1 | XP_007651513.1 | 8 - 9 | 1533 | 87 | |
Set Membership: |
|