Assay Details
| Gene Symbol: Slc30a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650376.1 | XP_007648566.1 | 4 - 5 | 558 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc30a2| Gene Symbol: Slc30a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650376.1 | XP_007648566.1 | 4 - 5 | 558 | 60 | |
Set Membership: |
|