Assay Details
| Gene Symbol: Hddc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652463.1 | XP_007650653.1 | 4 - 5 | 567 | 66 | |
| GenBank mRNA | JP038704.1 | - | 4 - 5 | 567 | 66 | |
More Information
Set Membership: |
|

Hddc2
Genomic Map